The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001212.3 | NP_001203.1 | 3 - 4 | 549 | 59 |
GenBank mRNA | AF275902.1 | - | 2 - 3 | 216 | 59 |
AK291516.1 | - | 3 - 4 | 509 | 59 | |
BC000435.2 | - | 3 - 4 | 503 | 59 | |
BC013731.1 | - | 3 - 4 | 492 | 59 | |
BT019898.1 | - | 3 - 4 | 471 | 59 | |
BT019899.1 | - | 3 - 4 | 471 | 59 | |
L04636.1 | - | 3 - 4 | 549 | 59 | |
X75913.1 | - | 3 - 4 | 549 | 59 |