The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001288705.1 | NP_001275634.1 | 15 - 16 | 2441 | 103 |
NM_005211.3 | NP_005202.2 | 16 - 17 | 2520 | 103 | |
NR_109969.1 | 14 - 15 | 2278 | 103 | ||
GenBank mRNA | AB208819.1 | - | 14 - 15 | 2228 | 103 |
BC047521.1 | - | 15 - 16 | 2423 | 103 | |
BX648599.1 | - | 14 - 15 | 2311 | 103 | |
X03663.1 | - | 16 - 17 | 2528 | 103 |