Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DNAJA3
Entrez Gene ID
9093
Gene Name
DnaJ heat shock protein family (Hsp40) member A3
Gene Aliases
HCA57, TID1, hTID-1
Chromosome Location
Chr.16: 4425805 - 4456775 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001135110.2 NP_001128582.1 9 - 10 1314 81
NM_001286516.1 NP_001273445.1 7 - 8 965 81
NM_005147.5 NP_005138.3 9 - 10 1314 81
GenBank mRNA AB209923.1 - 9 - 10 1233 81
AF061749.1 - 9 - 10 1268 81
AF411044.1 - 9 - 10 1281 81
AK001333.1 - 9 - 10 1254 81
AK223105.1 - 9 - 10 1254 81
AK295391.1 - 7 - 8 905 81
AK314218.1 - 9 - 10 1251 81
BC007225.1 - 9 - 10 1234 81
BC011855.2 - 9 - 10 1238 81
BC012343.1 - 9 - 10 1230 81
BC014062.2 - 9 - 10 1230 81
BC020248.1 - 5 - 6 563 81
BC030145.2 - 9 - 10 1230 81
BC032100.2 - 9 - 10 1252 81

View Full Product Details