The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_005401.4 | NP_005392.2 | 10 - 11 | 1587 | 82 |
XM_017001941.1 | XP_016857430.1 | 11 - 12 | 1674 | 82 | |
GenBank mRNA | AK291641.1 | - | 10 - 11 | 1519 | 82 |
AK298120.1 | - | 9 - 10 | 1180 | 82 | |
BC017300.2 | - | 10 - 11 | 1158 | 82 | |
BC101754.1 | - | 9 - 10 | 994 | 82 | |
BC104803.1 | - | 9 - 10 | 994 | 82 | |
HQ116786.1 | - | 8 - 9 | 846 | 82 | |
X82676.1 | - | 10 - 11 | 1128 | 82 |