The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_005401.4 | NP_005392.2 | 13 - 14 | 3200 | 108 |
XM_017001941.1 | XP_016857430.1 | 14 - 15 | 3287 | 108 | |
GenBank mRNA | AK291641.1 | - | 13 - 14 | 3132 | 108 |
AK298120.1 | - | 12 - 13 | 2793 | 108 | |
BC101754.1 | - | 12 - 13 | 2607 | 108 | |
BC104803.1 | - | 12 - 13 | 2607 | 108 | |
HQ116786.1 | - | 11 - 12 | 2459 | 108 | |
X82676.1 | - | 13 - 14 | 2741 | 108 |