The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_005401.4 | NP_005392.2 | 5 - 6 | 1166 | 115 |
XM_017001941.1 | XP_016857430.1 | 6 - 7 | 1253 | 115 | |
GenBank mRNA | AK291641.1 | - | 5 - 6 | 1098 | 115 |
AK298120.1 | - | 4 - 5 | 759 | 115 | |
BC017300.2 | - | 5 - 6 | 737 | 115 | |
BC101754.1 | - | 4 - 5 | 573 | 115 | |
BC104803.1 | - | 4 - 5 | 573 | 115 | |
HQ116786.1 | - | 4 - 5 | 513 | 115 | |
X82676.1 | - | 5 - 6 | 707 | 115 |