Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TSR1
Entrez Gene ID
55720
Gene Name
TSR1, ribosome maturation factor
Gene Aliases
-
Chromosome Location
Chr.17: 2322678 - 2337384 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_018128.4 NP_060598.3 5 - 6 1936 131
GenBank mRNA AB037822.1 - 5 - 6 2059 131
AK001396.1 - 3 - 4 578 131
AK026565.1 - 5 - 6 1009 131
BC019090.2 - 4 - 5 844 131
BC110851.1 - 5 - 6 1514 131
BC126110.1 - 5 - 6 1484 131

View Full Product Details