The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_018128.4 | NP_060598.3 | 6 - 7 | 2094 | 104 |
GenBank mRNA | AB037822.1 | - | 6 - 7 | 2217 | 104 |
AK001396.1 | - | 4 - 5 | 736 | 104 | |
AK026565.1 | - | 6 - 7 | 1167 | 104 | |
BC019090.2 | - | 5 - 6 | 1002 | 104 | |
BC110851.1 | - | 6 - 7 | 1672 | 104 | |
BC126110.1 | - | 6 - 7 | 1642 | 104 |