Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TSR1
Entrez Gene ID
55720
Gene Name
TSR1, ribosome maturation factor
Gene Aliases
-
Chromosome Location
Chr.17: 2322678 - 2337384 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_018128.4 NP_060598.3 3 - 4 1373 94
GenBank mRNA AB037822.1 - 3 - 4 1496 94
AK026565.1 - 3 - 4 446 94
BC019090.2 - 2 - 3 281 94
BC110851.1 - 3 - 4 951 94
BC126110.1 - 3 - 4 921 94

View Full Product Details