The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001618.3 | NP_001609.2 | 1 - 2 | 292 | 58 |
GenBank mRNA | AK225654.1 | - | 1 - 2 | 257 | 58 |
AK303340.1 | - | 1 - 2 | 282 | 58 | |
AK312339.1 | - | 1 - 2 | 208 | 58 | |
BC014206.2 | - | 1 - 2 | 271 | 58 | |
BC018620.1 | - | 1 - 2 | 252 | 58 | |
BC021045.1 | - | 1 - 2 | 126 | 58 | |
BC037545.1 | - | 1 - 2 | 265 | 58 | |
DB097441.1 | - | 1 - 2 | 292 | 58 | |
J03473.1 | - | 1 - 2 | 216 | 58 | |
M18112.1 | - | 1 - 2 | 260 | 58 | |
M32721.1 | - | 1 - 2 | 280 | 58 |