The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001618.3 | NP_001609.2 | 4 - 5 | 794 | 66 |
GenBank mRNA | AF401218.1 | - | 1 - 2 | 93 | 66 |
AK225654.1 | - | 4 - 5 | 759 | 66 | |
AK303340.1 | - | 4 - 5 | 721 | 66 | |
AK312339.1 | - | 4 - 5 | 710 | 66 | |
BC014206.2 | - | 4 - 5 | 773 | 66 | |
BC018620.1 | - | 4 - 5 | 754 | 66 | |
BC021045.1 | - | 4 - 5 | 628 | 66 | |
BC037545.1 | - | 4 - 5 | 767 | 66 | |
J03473.1 | - | 4 - 5 | 66 | ||
M18112.1 | - | 4 - 5 | 762 | 66 | |
M32721.1 | - | 4 - 5 | 782 | 66 |