The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001618.3 | NP_001609.2 | 9 - 10 | 1475 | 73 |
GenBank mRNA | AK225654.1 | - | 9 - 10 | 1440 | 73 |
AK303340.1 | - | 9 - 10 | 1402 | 73 | |
BC037545.1 | - | 9 - 10 | 1448 | 73 | |
J03473.1 | - | 9 - 10 | 1399 | 73 | |
M18112.1 | - | 9 - 10 | 1443 | 73 | |
M32721.1 | - | 9 - 10 | 1463 | 73 |