Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- MXRA7
- Entrez Gene ID
-
439921
- Gene Name
- matrix remodeling associated 7
- Gene Aliases
-
PS1TP1, TMAP1
- Chromosome Location
-
Chr.17: 76673650 - 76711005 on Build GRCh38
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
IMAGE Clone ID |
Transcript Location |
Amplicon Length |
GenBankmRNA |
BI964130.1 |
- |
1 |
408 |
|
Chr. 17 - 76674349 - 76674891 [-] on Build GRCh38 |
122 |
GenBankmRNA |
AL135722.1 |
- |
1 |
436 |
|
Chr. 17 - 76674298 - 76674919 [-] on Build GRCh38 |
122 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
uncharacterized LOC105274304 |
LOC105274304 |
|
|
Anti-sense |
Chr. - 76671928 - 76673658 on Build GRCh38 |
Downstream |
arginine demethylase and lysine hydroxylase |
JMJD6 |
PSR, PTDSR, PTDSR1 |
35.1kb
|
Sense |
Chr. - 76712830 - 76726799 on Build GRCh38 |
View Full Product Details