Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
BAHCC1
Entrez Gene ID
57597
Gene Name
BAH domain and coiled-coil containing 1
Gene Aliases
BAHD2
Chromosome Location
Chr.17: 81395431 - 81466332 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001291324.1 NP_001278253.1 7 - 8 2694 128
XM_011525063.2 XP_011523365.1 8 - 9 3086 128
XM_017024898.1 XP_016880387.1 7 - 8 2745 128
XM_017024899.1 XP_016880388.1 7 - 8 2744 128
GenBank mRNA AB040880.1 - 3 - 4 445 128

View Full Product Details