The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000861.3 | NP_000852.1 | 3 - 3 | 1822 | 80 |
NM_001098211.1 | NP_001091681.1 | 2 - 2 | 1743 | 80 | |
NM_001098212.1 | NP_001091682.1 | 2 - 2 | 1693 | 80 | |
NM_001098213.1 | NP_001091683.1 | 2 - 2 | 1973 | 80 | |
XM_011533652.1 | XP_011531954.1 | 4 - 4 | 2110 | 80 | |
XM_011533653.2 | XP_011531955.1 | 2 - 2 | 1939 | 80 | |
XM_017006283.1 | XP_016861772.1 | 2 - 2 | 1893 | 80 | |
XM_017006284.1 | XP_016861773.1 | 2 - 2 | 1932 | 80 | |
GenBank mRNA | AK289412.1 | - | 2 - 2 | 1973 | 80 |
AK292157.1 | - | 2 - 2 | 1690 | 80 | |
BC060802.1 | - | 3 - 3 | 1822 | 80 | |
Z34897.1 | - | 1 - 1 | 1631 | 80 |