The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_019022.3 | NP_061895.3 | 8 - 9 | 698 | 132 |
XM_005266714.2 | XP_005266771.1 | 7 - 8 | 643 | 132 | |
XM_011526033.2 | XP_011524335.1 | 6 - 7 | 561 | 132 | |
XM_011526034.2 | XP_011524336.1 | 8 - 9 | 690 | 132 | |
XM_011526035.2 | XP_011524337.1 | 8 - 9 | 690 | 132 | |
GenBank mRNA | AB058733.1 | - | 8 - 9 | 660 | 132 |
AK000800.1 | - | 2 - 3 | 138 | 132 | |
AK122715.1 | - | 8 - 9 | 690 | 132 | |
AK295551.1 | - | 7 - 8 | 558 | 132 | |
AK307269.1 | - | 7 - 8 | 571 | 132 | |
AK310389.1 | - | 8 - 9 | 880 | 132 | |
AL832341.1 | - | 8 - 9 | 646 | 132 | |
BC007690.2 | - | 7 - 8 | 489 | 132 | |
BC018981.2 | - | 7 - 8 | 489 | 132 | |
BC093792.1 | - | 8 - 9 | 658 | 132 | |
BC093794.1 | - | 8 - 9 | 659 | 132 | |
BC107422.1 | - | 8 - 9 | 694 | 132 | |
BC143400.1 | - | 7 - 8 | 613 | 132 | |
BX647846.1 | - | 8 - 9 | 707 | 132 |