The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_020922.4 | NP_065973.2 | 18 - 19 | 4320 | 78 |
XM_017029741.1 | XP_016885230.1 | 18 - 19 | 4259 | 78 | |
XM_017029742.1 | XP_016885231.1 | 18 - 19 | 4336 | 78 | |
XM_017029743.1 | XP_016885232.1 | 18 - 19 | 4251 | 78 | |
XM_017029744.1 | XP_016885233.1 | 18 - 19 | 4153 | 78 | |
XM_017029745.1 | XP_016885234.1 | 18 - 19 | 4258 | 78 | |
GenBank mRNA | AY082340.1 | - | 17 - 18 | 3881 | 78 |
AY352048.1 | - | 18 - 19 | 4320 | 78 |