The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NR_111971.1 | 11 - 12 | 1391 | 63 | |
GenBank mRNA | AK001594.1 | - | 9 - 10 | 1895 | 63 |
BF973758.1 | - | 2 - 3 | 164 | 63 |