Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TBX2
Entrez Gene ID
6909
Gene Name
T-box 2
Gene Aliases
-
Chromosome Location
Chr.17: 61399896 - 61409466 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_005994.3 NP_005985.3 4 - 5 1171 103
XM_011525159.1 XP_011523461.1 3 - 4 842 103
GenBank mRNA AB209378.1 - 4 - 5 702 103
AL832900.1 - 4 - 5 711 103
BC052566.1 - 4 - 5 1171 103
BC070054.1 - 4 - 5 1136 103
U28049.1 - 4 - 5 907 103

View Full Product Details