The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004750.4 | NP_004741.1 | 2 - 3 | 594 | 72 |
XM_011528422.2 | XP_011526724.1 | 2 - 3 | 676 | 72 | |
XM_011528423.2 | XP_011526725.1 | 2 - 3 | 400 | 72 | |
XM_011528424.2 | XP_011526726.1 | 2 - 3 | 668 | 72 | |
GenBank mRNA | AF059293.1 | - | 2 - 3 | 518 | 72 |
AF073515.1 | - | 2 - 3 | 603 | 72 | |
AF178684.1 | - | 2 - 3 | 451 | 72 | |
AY358291.1 | - | 2 - 3 | 530 | 72 | |
BC044634.1 | - | 2 - 3 | 527 | 72 |