The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001136191.2 | NP_001129663.1 | 3 - 4 | 399 | 152 |
NM_015493.6 | NP_056308.3 | 1 - 2 | 117 | 152 | |
GenBank mRNA | AB040951.2 | - | 2 - 3 | 351 | 152 |
AB284125.1 | - | 4 - 5 | 582 | 152 | |
AK000011.1 | - | 2 - 3 | 214 | 152 | |
AK225038.1 | - | 2 - 3 | 202 | 152 | |
AL701379.1 | - | 3 - 4 | 408 | 152 | |
BC082762.1 | - | 3 - 4 | 353 | 152 | |
BC098105.1 | - | 2 - 3 | 151 | 152 | |
BC098286.1 | - | 2 - 3 | 151 | 152 | |
BC098312.1 | - | 2 - 3 | 151 | 152 | |
CN306249.1 | - | 2 - 3 | 548 | 152 | |
DA758311.1 | - | 3 - 4 | 446 | 152 | |
DA991654.1 | - | 3 - 4 | 433 | 152 | |
DB282630.1 | - | 2 - 3 | 229 | 152 |