The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001010972.1 | NP_001010972.1 | 5 - 6 | 1182 | 73 |
NM_003461.4 | NP_003452.1 | 5 - 6 | 1185 | 73 | |
XM_011516569.2 | XP_011514871.2 | 4 - 5 | 1080 | 73 | |
XM_017012587.1 | XP_016868076.1 | 5 - 6 | 1275 | 73 | |
GenBank mRNA | AK130138.1 | - | 1 - 2 | 475 | 73 |
AK298925.1 | - | 3 - 4 | 931 | 73 | |
AK299005.1 | - | 4 - 5 | 901 | 73 | |
AK302309.1 | - | 4 - 5 | 993 | 73 | |
AK316227.1 | - | 4 - 5 | 877 | 73 | |
BC002323.2 | - | 5 - 6 | 1054 | 73 | |
BC008743.2 | - | 5 - 6 | 1149 | 73 | |
BC009360.2 | - | 5 - 6 | 1094 | 73 | |
BC010031.2 | - | 5 - 6 | 1082 | 73 | |
BC017183.2 | - | 4 - 5 | 711 | 73 | |
CR457431.1 | - | 4 - 5 | 1027 | 73 | |
U15158.1 | - | 3 - 4 | 790 | 73 | |
X94991.1 | - | 4 - 5 | 1037 | 73 | |
X95735.1 | - | 5 - 6 | 1098 | 73 |