Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SIN3A
Entrez Gene ID
25942
Gene Name
SIN3 transcription regulator family member A
Gene Aliases
-
Chromosome Location
Chr.15: 75369379 - 75455819 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001145357.1 NP_001138829.1 10 - 11 1706 92
NM_001145358.1 NP_001138830.1 10 - 11 1881 92
NM_015477.2 NP_056292.1 10 - 11 1783 92
XM_006720465.3 XP_006720528.1 10 - 11 1589 92
XM_006720466.3 XP_006720529.1 11 - 12 1979 92
XM_006720467.3 XP_006720530.1 12 - 13 1797 92
GenBank mRNA AF418569.1 - 10 - 11 1783 92
AK027559.1 - 10 - 11 1783 92
AK096477.1 - 10 - 11 1832 92
AL832463.1 - 10 - 11 1707 92
AY044430.1 - 9 - 10 1569 92
BC137098.1 - 10 - 11 1634 92
BC137099.1 - 10 - 11 1634 92
BX649162.1 - 10 - 11 1820 92

View Full Product Details