The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_015378.3 | NP_056193.2 | 51 - 52 | 10430 | 74 |
NM_018156.3 | NP_060626.2 | 50 - 51 | 10355 | 74 | |
GenBank mRNA | AB007922.3 | - | 33 - 34 | 6737 | 74 |
AJ608774.1 | - | 51 - 52 | 10402 | 74 | |
AJ608775.1 | - | 50 - 51 | 10327 | 74 | |
AK057675.1 | - | 9 - 10 | 1399 | 74 | |
AK125118.1 | - | 25 - 26 | 4275 | 74 | |
AL833320.1 | - | - | 756 | 74 | |
BC028115.1 | - | 9 - 10 | 1346 | 74 | |
BX641035.1 | - | 25 - 26 | 3919 | 74 |