The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001018000.3 | NP_001018000.1 | 1 - 2 | 444 | 81 |
XM_011541075.2 | XP_011539377.1 | 1 - 2 | 425 | 81 | |
GenBank mRNA | AY505120.1 | - | 1 - 2 | 444 | 81 |
BC035501.2 | - | 1 - 2 | 443 | 81 |