Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DYNC1I2
Entrez Gene ID
1781
Gene Name
dynein cytoplasmic 1 intermediate chain 2
Gene Aliases
DIC74, DNCI2, IC2
Chromosome Location
Chr.2: 171687409 - 171750158 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001271785.1 NP_001258714.1 3 - 4 438 85
NM_001378.2 NP_001369.1 3 - 4 455 85
GenBank mRNA AK055491.1 - 3 - 4 392 85
AK096579.1 - 3 - 4 356 85

View Full Product Details