The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001271790.1 | NP_001258719.1 | 15 - 16 | 1946 | 81 |
NM_001320884.1 | NP_001307813.1 | 15 - 16 | 1929 | 81 | |
XM_006712347.3 | XP_006712410.1 | 16 - 17 | 1930 | 81 | |
XM_017003525.1 | XP_016859014.1 | 16 - 17 | 1965 | 81 | |
GenBank mRNA | AY037160.1 | - | 15 - 16 | 1854 | 81 |