The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001319035.1 | NP_001305964.1 | 19 - 20 | 2823 | 86 |
NM_007110.4 | NP_009041.2 | 21 - 22 | 3147 | 86 | |
XM_005268027.4 | XP_005268084.1 | 21 - 22 | 3287 | 86 | |
XM_011537110.2 | XP_011535412.1 | 13 - 14 | 1632 | 86 | |
GenBank mRNA | AB209669.1 | - | 9 - 10 | 1154 | 86 |
BC126107.1 | - | 21 - 22 | 3147 | 86 | |
BC143812.1 | - | 20 - 21 | 3146 | 86 | |
U86136.2 | - | 21 - 22 | 3147 | 86 |