The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001319035.1 | NP_001305964.1 | 3 - 4 | 782 | 82 |
NM_007110.4 | NP_009041.2 | 3 - 4 | 782 | 82 | |
XM_005268027.4 | XP_005268084.1 | 3 - 4 | 922 | 82 | |
GenBank mRNA | BC126107.1 | - | 3 - 4 | 782 | 82 |
BC143812.1 | - | 2 - 3 | 781 | 82 | |
U86136.2 | - | 3 - 4 | 782 | 82 |