Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CPT1A
Entrez Gene ID
1374
Gene Name
carnitine palmitoyltransferase 1A
Gene Aliases
CPT1, CPT1-L, L-CPT1
Chromosome Location
Chr.11: 68754620 - 68844410 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001031847.2 NP_001027017.1 16 - 17 2200 93
NM_001876.3 NP_001867.2 16 - 17 2200 93
XM_005273762.2 XP_005273819.1 16 - 17 2176 93
XM_005273763.1 XP_005273820.1 16 - 17 2176 93
XM_017017220.1 XP_016872709.1 16 - 17 2309 93
GenBank mRNA AK025307.1 - 11 - 12 1486 93
AK172798.1 - 16 - 17 2188 93
AK314301.1 - 16 - 17 2139 93
BC000185.1 - 16 - 17 2075 93
BT009791.1 - 15 - 16 2030 93
L39211.1 - 16 - 17 2106 93

View Full Product Details