Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CPT1A
Entrez Gene ID
1374
Gene Name
carnitine palmitoyltransferase 1A
Gene Aliases
CPT1, CPT1-L, L-CPT1
Chromosome Location
Chr.11: 68754620 - 68844410 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001031847.2 NP_001027017.1 17 - 18 2311 86
NM_001876.3 NP_001867.2 17 - 18 2311 86
XM_005273762.2 XP_005273819.1 17 - 18 2287 86
XM_005273763.1 XP_005273820.1 17 - 18 2287 86
XM_017017220.1 XP_016872709.1 17 - 18 2420 86
GenBank mRNA AK025307.1 - 12 - 13 1597 86
AK172798.1 - 17 - 18 2299 86
AK314301.1 - 17 - 18 2250 86
BC000185.1 - 17 - 18 2186 86
BT009791.1 - 16 - 17 2141 86
L39211.1 - 17 - 18 2217 86

View Full Product Details