Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CPT1A
Entrez Gene ID
1374
Gene Name
carnitine palmitoyltransferase 1A
Gene Aliases
CPT1, CPT1-L, L-CPT1
Chromosome Location
Chr.11: 68754620 - 68844410 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001031847.2 NP_001027017.1 1 - 2 160 108
NM_001876.3 NP_001867.2 1 - 2 160 108
GenBank mRNA AK172798.1 - 1 - 2 146 108
AK314301.1 - 1 - 2 99 108
DB063502.1 - 1 - 2 160 108

View Full Product Details