The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_018109.3 | NP_060579.3 | 7 - 8 | 1368 | 141 |
GenBank mRNA | AB194709.1 | - | 7 - 8 | 1305 | 141 |
AK001348.1 | - | 7 - 8 | 1309 | 141 | |
AK022188.1 | - | 7 - 8 | 1348 | 141 | |
AY364242.1 | - | 7 - 8 | 1305 | 141 | |
BC061703.1 | - | 7 - 8 | 1363 | 141 |