The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001308243.1 | NP_001295172.1 | 3 - 4 | 585 | 65 |
GenBank mRNA | AF008556.1 | - | 2 - 3 | 278 | 65 |
CR996090.1 | - | 3 - 4 | 494 | 65 |