Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DAZAP1
Entrez Gene ID
26528
Gene Name
DAZ associated protein 1
Gene Aliases
-
Chromosome Location
Chr.19: 1407569 - 1435687 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005259536.3 XP_005259593.1 10 - 11 1132 59
XM_011527905.2 XP_011526207.1 11 - 12 2527 59
XM_011527907.2 XP_011526209.1 11 - 12 2533 59
XM_017026581.1 XP_016882070.1 11 - 12 1249 59
GenBank mRNA AK125032.1 - 9 - 10 2467 59
KU178594.1 - 3 - 4 348 59

View Full Product Details