Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CHRM1
Entrez Gene ID
1128
Gene Name
cholinergic receptor muscarinic 1
Gene Aliases
HM1, M1, M1R
Chromosome Location
Chr.11: 62908675 - 62921861 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_000738.2 NP_000729.2 1 - 2 326 63
GenBank mRNA AJ006520.1 - 1 - 2 342 63
BC007740.2 - 1 - 2 47 63
BC022984.1 - 1 - 2 326 63

View Full Product Details