Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CHRM1
Entrez Gene ID
1128
Gene Name
cholinergic receptor muscarinic 1
Gene Aliases
HM1, M1, M1R
Chromosome Location
Chr.11: 62908675 - 62921861 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_000738.2 NP_000729.2 2 - 2 2488 63
XM_011544742.2 XP_011543044.1 2 - 2 2189 63
GenBank mRNA BC007740.2 - 2 - 2 2209 63
BC022984.1 - 2 - 2 2488 63

View Full Product Details