Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SLC25A36
Entrez Gene ID
55186
Gene Name
solute carrier family 25 member 36
Gene Aliases
PNC2
Chromosome Location
Chr.3: 140941820 - 140979943 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001104647.1 NP_001098117.1 4 - 5 622 152
NM_018155.2 NP_060625.2 4 - 5 622 152
GenBank mRNA AK001480.1 - 4 - 5 596 152
AK296785.1 - 3 - 4 463 152
BC014064.1 - 4 - 5 555 152

View Full Product Details