The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001256378.1 | NP_001243307.1 | 12 - 13 | 1712 | 84 |
NM_001256379.1 | NP_001243308.1 | 12 - 13 | 1522 | 84 | |
NM_024834.3 | NP_079110.1 | 12 - 13 | 1718 | 84 | |
XM_005270157.4 | XP_005270214.1 | 12 - 13 | 1800 | 84 | |
XM_011540170.1 | XP_011538472.1 | 12 - 13 | 1559 | 84 | |
XM_017016663.1 | XP_016872152.1 | 12 - 13 | 1794 | 84 | |
XM_017016664.1 | XP_016872153.1 | 12 - 13 | 1553 | 84 | |
GenBank mRNA | AK023143.1 | - | 12 - 13 | 1589 | 84 |
AK094075.1 | - | 12 - 13 | 1516 | 84 | |
BC000935.2 | - | 3 - 4 | 404 | 84 | |
BC004183.2 | - | 12 - 13 | 1678 | 84 | |
BC007219.1 | - | 12 - 13 | 1678 | 84 | |
CR457299.1 | - | 12 - 13 | 1413 | 84 | |
KU178788.1 | - | 12 - 13 | 1413 | 84 | |
KU178789.1 | - | 12 - 13 | 1382 | 84 |