The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_007190.3 | NP_009121.1 | 12 - 13 | 2323 | 87 |
NR_037771.1 | 11 - 12 | 1790 | 87 | ||
XM_005269469.2 | XP_005269526.1 | 12 - 13 | 2278 | 87 | |
XM_005269470.2 | XP_005269527.1 | 12 - 13 | 2349 | 87 | |
XM_017015610.1 | XP_016871099.1 | 12 - 13 | 2349 | 87 | |
GenBank mRNA | AB019435.1 | - | 12 - 13 | 2185 | 87 |
AF116723.1 | - | 12 - 13 | 2145 | 87 | |
AK021846.1 | - | 8 - 9 | 1538 | 87 | |
AK023238.1 | - | 3 - 4 | 352 | 87 | |
AK301517.1 | - | 11 - 12 | 1633 | 87 | |
AK314592.1 | - | 12 - 13 | 2180 | 87 | |
BC063800.1 | - | 12 - 13 | 2163 | 87 | |
BX649119.1 | - | 12 - 13 | 2118 | 87 |