Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PLEKHS1
Entrez Gene ID
79949
Gene Name
pleckstrin homology domain containing S1
Gene Aliases
C10orf81, HEL185, bA211N11.2
Chromosome Location
Chr.10: 113751262 - 113782433 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001193434.1 NP_001180363.1 9 - 10 1202 87
NM_001193435.1 NP_001180364.1 7 - 8 1006 87
NM_024889.4 NP_079165.3 9 - 10 1102 87
NM_182601.1 NP_872407.1 8 - 9 1319 87
XM_005270162.2 XP_005270219.1 9 - 10 1241 87
XM_005270163.2 XP_005270220.1 9 - 10 1241 87
XM_011540171.1 XP_011538473.1 9 - 10 1241 87
XM_011540172.1 XP_011538474.1 10 - 11 1009 87
XM_011540173.1 XP_011538475.1 9 - 10 1241 87
XM_011540175.1 XP_011538477.1 9 - 10 1180 87
XM_011540176.1 XP_011538478.1 7 - 8 778 87
XM_017016666.1 XP_016872155.1 9 - 10 1241 87
GenBank mRNA AK027190.1 - 2 - 3 165 87
AK126354.1 - 7 - 8 778 87
AK128811.1 - 9 - 10 1102 87
AK292867.1 - 9 - 10 974 87
BC036365.1 - 3 - 4 256 87
BC037870.1 - 8 - 9 1324 87
FJ236306.1 - 8 - 9 1324 87

View Full Product Details