Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PLEKHS1
Entrez Gene ID
79949
Gene Name
pleckstrin homology domain containing S1
Gene Aliases
C10orf81, HEL185, bA211N11.2
Chromosome Location
Chr.10: 113751262 - 113782433 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001193434.1 NP_001180363.1 12 - 13 1606 68
NM_001193435.1 NP_001180364.1 10 - 11 1410 68
XM_005270162.2 XP_005270219.1 12 - 13 1687 68
XM_005270163.2 XP_005270220.1 12 - 13 1645 68
GenBank mRNA AK027190.1 - 5 - 6 569 68
AK126354.1 - 10 - 11 1182 68
AK292867.1 - 12 - 13 1378 68
AK296132.1 - 2 - 3 542 68
BC036365.1 - 6 - 7 702 68

View Full Product Details