The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_030787.3 | NP_110414.1 | 8 - 9 | 1455 | 97 |
XM_011510020.2 | XP_011508322.1 | 8 - 9 | 1362 | 97 | |
GenBank mRNA | AF295327.1 | - | 8 - 9 | 1420 | 97 |
BC111773.1 | - | 8 - 9 | 1420 | 97 |