Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SORBS1
Entrez Gene ID
10580
Gene Name
sorbin and SH3 domain containing 1
Gene Aliases
CAP, FLAF2, R85FL, SH3D5, SH3P12, SORB1
Chromosome Location
Chr.10: 95311773 - 95561420 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001034957.1 NP_001030129.1 4 - 5 364 98
NM_001290295.1 NP_001277224.1 7 - 8 578 98
NM_001290297.1 NP_001277226.1 5 - 6 391 98
NM_001290298.1 NP_001277227.1 5 - 6 460 98
NM_006434.2 NP_006425.2 4 - 5 364 98
NM_024991.2 NP_079267.1 8 - 9 674 98
XM_006717593.1 XP_006717656.1 6 - 7 529 98
XM_017015504.1 XP_016870993.1 8 - 9 627 98
XM_017015506.1 XP_016870995.1 8 - 9 627 98
XM_017015508.1 XP_016870997.1 8 - 9 627 98
XM_017015509.1 XP_016870998.1 7 - 8 556 98
XM_017015511.1 XP_016871000.1 8 - 9 627 98
XM_017015512.1 XP_016871001.1 6 - 7 529 98
XM_017015513.1 XP_016871002.1 6 - 7 529 98
XM_017015514.1 XP_016871003.1 6 - 7 529 98
XM_017015525.1 XP_016871014.1 7 - 8 556 98
XM_017015526.1 XP_016871015.1 8 - 9 627 98
XM_017015527.1 XP_016871016.1 7 - 8 556 98
XM_017015528.1 XP_016871017.1 7 - 8 600 98
XM_017015529.1 XP_016871018.1 8 - 9 627 98
XM_017015530.1 XP_016871019.1 8 - 9 627 98
XM_017015531.1 XP_016871020.1 7 - 8 556 98
XM_017015532.1 XP_016871021.1 7 - 8 600 98
XM_017015533.1 XP_016871022.1 6 - 7 529 98
XM_017015534.1 XP_016871023.1 8 - 9 627 98
XM_017015535.1 XP_016871024.1 7 - 8 556 98
XM_017015536.1 XP_016871025.1 7 - 8 556 98
XM_017015537.1 XP_016871026.1 7 - 8 600 98
XM_017015538.1 XP_016871027.1 7 - 8 600 98
XM_017015539.1 XP_016871028.1 6 - 7 529 98
XM_017015540.1 XP_016871029.1 6 - 7 529 98
GenBank mRNA AB037717.1 - 8 - 9 606 98
AF330623.1 - 4 - 5 346 98
AJ489942.1 - 4 - 5 415 98
AK300407.1 - 7 - 8 531 98
BC152463.1 - 8 - 9 606 98

View Full Product Details