The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002703.4 | NP_002694.3 | 6 - 7 | 889 | 76 |
XM_011534387.1 | XP_011532689.1 | 6 - 7 | 666 | 76 | |
GenBank mRNA | AK222759.1 | - | 6 - 7 | 76 | |
AK290942.1 | - | 6 - 7 | 744 | 76 | |
AK312297.1 | - | 6 - 7 | 865 | 76 | |
BC004200.2 | - | 6 - 7 | 761 | 76 | |
D13757.1 | - | 6 - 7 | 851 | 76 | |
U00238.1 | - | 6 - 7 | 780 | 76 |