The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001206997.1 | NP_001193926.1 | 6 - 7 | 710 | 160 |
NM_152770.2 | NP_689983.2 | 5 - 6 | 659 | 160 | |
XM_017007972.1 | XP_016863461.1 | 6 - 7 | 811 | 160 | |
XM_017007975.1 | XP_016863464.1 | 4 - 5 | 799 | 160 | |
XM_017007978.1 | XP_016863467.1 | 4 - 5 | 676 | 160 | |
GenBank mRNA | AI650567.1 | - | 1 - 2 | 160 | |
AY349359.1 | - | 6 - 7 | 661 | 160 | |
BC034296.1 | - | 5 - 6 | 659 | 160 | |
DB522959.1 | - | 2 - 3 | 218 | 160 |