Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RWDD3
Entrez Gene ID
25950
Gene Name
RWD domain containing 3
Gene Aliases
RSUME
Chromosome Location
Chr.1: 95234155 - 95247225 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NR_103483.1 2 - 3 672 81
GenBank mRNA BC010936.2 - 2 - 3 608 81

View Full Product Details