The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001317958.1 | NP_001304887.1 | 6 - 7 | 1053 | 84 |
NM_001317959.1 | NP_001304888.1 | 7 - 8 | 1146 | 84 | |
NM_152572.2 | NP_689785.1 | 6 - 7 | 1046 | 84 | |
XM_005272169.2 | XP_005272226.1 | 6 - 7 | 585 | 84 | |
XM_006716965.2 | XP_006717028.1 | 4 - 5 | 358 | 84 | |
XM_011518277.2 | XP_011516579.1 | 6 - 7 | 585 | 84 | |
XM_011518278.2 | XP_011516580.1 | 5 - 6 | 426 | 84 | |
GenBank mRNA | AK057266.1 | - | 6 - 7 | 519 | 84 |
AK093446.1 | - | 6 - 7 | 561 | 84 | |
AK292186.1 | - | 6 - 7 | 531 | 84 | |
BC034776.1 | - | 6 - 7 | 543 | 84 | |
BC050576.1 | - | 6 - 7 | 1046 | 84 |