Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MKX
Entrez Gene ID
283078
Gene Name
mohawk homeobox
Gene Aliases
C10orf48, IFRX, IRXL1
Chromosome Location
Chr.10: 27672874 - 27745849 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001242702.1 NP_001229631.1 5 - 6 1063 86
NM_173576.2 NP_775847.2 5 - 6 1066 86
XM_017016105.1 XP_016871594.1 5 - 6 1103 86
XM_017016106.1 XP_016871595.1 5 - 6 1294 86
GenBank mRNA AK125339.1 - 5 - 6 1068 86
BC036207.1 - 5 - 6 86

View Full Product Details