Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DACH2
Entrez Gene ID
117154
Gene Name
dachshund family transcription factor 2
Gene Aliases
-
Chromosome Location
Chr.X: 86148451 - 86832602 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001139514.1 NP_001132986.1 5 - 6 1241 66
NM_001139515.1 NP_001132987.1 6 - 7 666 66
NM_053281.3 NP_444511.1 6 - 7 1280 66
XM_011530846.2 XP_011529148.1 6 - 7 1279 66
XM_011530847.2 XP_011529149.1 6 - 7 1279 66
XM_011530848.2 XP_011529150.1 6 - 7 1279 66
XM_017029254.1 XP_016884743.1 5 - 6 1240 66
XM_017029255.1 XP_016884744.1 5 - 6 1240 66
XM_017029256.1 XP_016884745.1 5 - 6 1240 66
GenBank mRNA AF428101.1 - 6 - 7 1110 66
AK055953.1 - 5 - 6 627 66
AK091842.1 - 5 - 6 1234 66
AK290596.1 - 6 - 7 1280 66
AK294498.1 - 1 - 2 75 66
AK295648.1 - 1 - 2 76 66
AK298787.1 - 6 - 7 666 66
BC114950.1 - 6 - 7 1189 66

View Full Product Details